Merck
  • α7 nicotinic acetylcholine receptor signaling modulates the inflammatory phenotype of fetal brain microglia: first evidence of interference by iron homeostasis.
α7 nicotinic acetylcholine receptor signaling modulates the inflammatory phenotype of fetal brain microglia: first evidence of interference by iron homeostasis.
Scientific reports (2017-09-08)
M Cortes, M Cao, H L Liu, C S Moore, L D Durosier, P Burns, G Fecteau, A Desrochers, L B Barreiro, J P Antel, M G Frasch
ABSTRACT
Neuroinflammation in utero may result in life-long neurological disabilities. Microglia play a pivotal role, but the mechanisms are poorly understood. No early postnatal treatment strategies exist to enhance neuroprotective potential of microglia. We hypothesized that agonism on α7 nicotinic acetylcholine receptor (α7nAChR) in fetal microglia will augment their neuroprotective transcriptome profile, while the antagonistic stimulation of α7nAChR will achieve the opposite. Using an in vivo - in vitro model of developmental programming of neuroinflammation induced by lipopolysaccharide (LPS), we validated this hypothesis in primary fetal sheep microglia cultures re-exposed to LPS in presence of a selective α7nAChR agonist or antagonist. Our RNAseq and protein level findings show that a pro-inflammatory microglial phenotype acquired in vitro by LPS stimulation is reversed with α7nAChR agonistic stimulation. Conversely, antagonistic α7nAChR stimulation potentiates the pro-inflammatory microglial phenotype. Surprisingly, under conditions of LPS double-hit an interference of a postulated α7nAChR - ferroportin signaling pathway may impede this mechanism. These results suggest a therapeutic potential of α7nAChR agonists in early re-programming of microglia in neonates exposed to in utero inflammation via an endogenous cerebral cholinergic anti-inflammatory pathway. Future studies will assess the role of interactions between inflammation-triggered microglial iron sequestering and α7nAChR signaling in neurodevelopment.
Sigma-Aldrich
1,2-Dilinoleoyl-3-palmitoyl-rac-glycerol, ≥95% (TLC), liquid
Pricing and availability is not currently available.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O127:B8, purified by ion-exchange chromatography, TLR ligand tested
Pricing and availability is not currently available.