Merck
  • Fabrication and characterization of gold nanoparticle-loaded TiO2 nanotube arrays for medical implants.
Fabrication and characterization of gold nanoparticle-loaded TiO2 nanotube arrays for medical implants.
Journal of materials science. Materials in medicine (2015-12-26)
Yu Bai, Yulong Bai, Cunyang Wang, Jingjun Gao, Wen Ma
ABSTRACT
Au nanoparticles (AuNPs) are successfully assembled on TiO2 nanotube (TN) arrays through electrochemical deposition technology to improve the surface characteristics of TN arrays as an implant material. The loading amount of AuNPs can be controlled by adjusting the deposition time of electrochemical deposition. The effect of the amount of the loaded AuNPs on surface roughness and surface energy is systematically investigated on the basis of various characterizations. Results show that the increase in the loading amount of AuNPs on the TN arrays can increase surface roughness and decrease surface energy. Potentiodynamic polarization tests indicate that AuNP-modified TNs possess a higher corrosion resistance than unmodified TNs. Corrosion resistance increases as the amount of the loaded AuNP increases. In vitro cell culture tests are performed on the basis of cell morphology observations and MTT assays. Osteoblast cell adhesion and proliferation ability on the AuNP-modified TN surface are greater than those on the unmodified TN surface. The sample fabricated at the deposition time of 90 s exhibits an optimum cell performance. This work can provide a new platform to develop the surface chemistry of TN arrays and to fabricate titanium-based implant materials to enhance bioactivity.
Product Description
SAFC
Hanks′ Balanced Salts, Modified, without calcium chloride, magnesium sulfate and sodium bicarbonate, powder, suitable for cell culture
Pricing and availability is not currently available.