Merck
  • Prolonged Ketamine Effects in Sp4 Hypomorphic Mice: Mimicking Phenotypes of Schizophrenia.
Prolonged Ketamine Effects in Sp4 Hypomorphic Mice: Mimicking Phenotypes of Schizophrenia.
PloS one (2013-07-05)
Baohu Ji, Xin Wang, Antonio Pinto-Duarte, Minjung Kim, Sorana Caldwell, Jared W Young, Margarita M Behrens, Terrence J Sejnowski, Mark A Geyer, Xianjin Zhou
ABSTRACT
It has been well established that schizophrenia patients display impaired NMDA receptor (NMDAR) functions as well as exacerbation of symptoms in response to NMDAR antagonists. Abnormal NMDAR signaling presumably contributes to cognitive deficits which substantially contribute to functional disability in schizophrenia. Establishing a mouse genetic model will help investigate molecular mechanisms of hypoglutmatergic neurotransmission in schizophrenia. Here, we examined the responses of Sp4 hypomorphic mice to NMDAR antagonists in electroencephalography and various behavioral paradigms. Sp4 hypomorphic mice, previously reported to have reduced NMDAR1 expression and LTP deficit in hippocampal CA1, displayed increased sensitivity and prolonged responses to NMDAR antagonists. Molecular studies demonstrated reduced expression of glutamic acid decarboxylase 67 (GAD67) in both cortex and hippocampus, consistent with abnormal gamma oscillations in Sp4 hypomorphic mice. On the other hand, human SP4 gene was reported to be deleted in schizophrenia. Several human genetic studies suggested the association of SP4 gene with schizophrenia and other psychiatric disorders. Therefore, elucidation of the Sp4 molecular pathway in Sp4 hypomorphic mice may provide novel insights to our understanding of abnormal NMDAR signaling in schizophrenia.
Sigma-Aldrich
Monoclonal Anti-Glutamic Acid Decarboxylase 67 (GAD67) antibody produced in mouse, clone K-87, purified antibody
Sigma-Aldrich
Monoclonal Anti-Parvalbumin antibody produced in mouse, clone PARV-19, ascites fluid