Merck
Molecular cloning and expression of human hepatocyte growth factor.
Nature (1989-11-23)
T Nakamura, T Nishizawa, M Hagiya, T Seki, M Shimonishi, A Sugimura, K Tashiro, S Shimizu
ABSTRACT
Hepatocyte growth factor (HGF) is the most potent mitogen for mature parenchymal hepatocytes in primary culture, and seems to be a hepatotrophic factor that acts as a trigger for liver regeneration after partial hepatectomy and liver injury. The partial purification and characterization of HGF have been reported. We have demonstrated that pure HGF from rat platelets is a new growth factor effective at concentrations as low as 1 ng ml-1. The effects of HGF and epidermal growth factor (EGF) are additive. The activity of HGF is not species-specific, although it does not stimulate growth in Swiss 3T3 fibroblasts. HGF has a relative molecular mass (Mr) of 82,000 and is a heterodimer composed of a large alpha-subunit of Mr 69,000 and a small beta-subunit of Mr 34,000. Here we report the amino-acid sequence of human HGF determined by complementary DNA cloning and the expression of biologically active human HGF from COS-1 cells transfected with cloned cDNA. The nucleotide sequence of the human HGF cDNA reveals that both alpha- and beta-chains are contained in a single open reading frame coding for a pre-pro precursor protein of 728 amino acids.
Sigma-Aldrich
Hepatocyte Growth Factor human, HGF, recombinant, expressed in NSO cells, suitable for cell culture
Sigma-Aldrich
Hepatocyte Growth Factor human, HGF, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture