Merck
  • Application of circular dichroism and magnetic circular dichroism for assessing biopharmaceuticals formulations photo-stability and small ligands binding properties.
Application of circular dichroism and magnetic circular dichroism for assessing biopharmaceuticals formulations photo-stability and small ligands binding properties.
International journal of pharmaceutics (2015-01-18)
Edoardo Longo, Rohanah Hussain, Giuliano Siligardi
ABSTRACT
Synchrotron radiation circular dichroism (SRCD) is a powerful tool for photo-stability assessment of proteins. Recently our research has been interested in applying SRCD to develop screening methodologies for accelerated photo-stability assessment of monoclonal antibody formulations. Despite it was proven to be reliable and applicable within a wide range of salts and excipients containing solutions, the presence of far-UV (<260nm) strong absorbing species (e.g., sodium chloride, histidine, arginine) in common formulations completely prevent the analysis. Herein, we propose a new method based on CD coupled with magnetic CD (MCD) to address the problem and offer an additional versatile tool for monitoring the photo-stability. This is done by assessing the stability of the samples by looking at the near-UV band, as well as giving insights in the denaturation mechanism. We applied this method to four mAbs formulations and correlated the results with dynamic light scattering data. Finally, we applied MCD in ligand interaction to key proteins such as lysozyme, comparing the human with the hen enzyme in the binding of N,N',N''-triacetylchitotriose.
Sigma-Aldrich
Myoglobin from equine skeletal muscle, 95-100%, essentially salt-free, lyophilized powder
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥39,000 units/mg protein
Sigma-Aldrich
Lysozyme human, Lysobac, recombinant, expressed in rice, ≥100,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), Type VI, lyophilized powder
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially IgG free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from human serum, lyophilized powder, Fatty acid free, Globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
α-Lactalbumin from bovine milk, Type III, calcium depleted, ≥85% (PAGE), lyophilized powder