Merck
  • Ecological risk of estrogenic endocrine disrupting chemicals in sewage plant effluent and reclaimed water.
Ecological risk of estrogenic endocrine disrupting chemicals in sewage plant effluent and reclaimed water.
Environmental pollution (Barking, Essex : 1987) (2013-06-06)
Yan Sun, Huang Huang, Ying Sun, Chao Wang, Xiao-Lei Shi, Hong-Ying Hu, Takashi Kameya, Koichi Fujie
ABSTRACT
The long-term ecological risk of micropollutants, especially endocrine disrupting chemicals (EDCs) has threatened reclaimed water quality. In this study, estrogenic activity and ecological risk of eight typical estrogenic EDCs in effluents from sewage plants were evaluated. The estrogenic activity analysis showed that steroidal estrogens had the highest estrogenic activity (ranged from 10(-1) to 10(3) ng-E2/L), phenolic compounds showed weaker estrogenic activity (mainly ranged from 10(-3) to 10 ng-E2/L), and phthalate esters were negligible. The ecological risk of the estrogenic EDCs which was characterized by risk quotient ranged from 10(-4) to 10(3), with an order in descending: steroids estrogens, phenolic compounds and phthalate esters. The eight estrogenic EDCs were scored and sorted based on the comparison of the estrogenic activity and the ecological risk, suggesting that 17α-ethynylestradiol (EE2), estrone (E1) and estradiol (E2) should be the priority EDCs to control in municipal sewage plants.
Product Description
Ethinylestradiol for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
17α-Ethynylestradiol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Estrone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Estrone, European Pharmacopoeia (EP) Reference Standard
Ethinylestradiol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
17α-Ethynylestradiol, ≥98%
Sigma-Aldrich
Estrone, ≥99%