Merck
  • Biological activities of juvenile hormone III skipped bisepoxide in last instar nymphs and adults of a stink bug, Plautia stali.
Biological activities of juvenile hormone III skipped bisepoxide in last instar nymphs and adults of a stink bug, Plautia stali.
Journal of insect physiology (2010-10-26)
Toyomi Kotaki, Tetsuro Shinada, Kanako Kaihara, Yasufumi Ohfune, Hideharu Numata
ABSTRACT
Juvenile hormone III skipped bisepoxide (JHSB(3)), methyl (2R,3S,10R)-2,3;10,11-bisepoxyfarnesoate was recently determined as a novel juvenile hormone (JH) in a stink bug, Plautia stali. To further confirm the biological function of JHSB(3) in this insect, its juvenilizing, reproduction-stimulating and diapause-terminating activities and the presence in the hemolymph were examined. Topical application of JHSB(3) to last instar nymphs inhibited their metamorphosis in a dose-dependent fashion. In allatectomized and diapausing adults, JHSB(3) application exerted stimulatory effects on the development of ovaries and ectadenia in females and males, respectively. JHSB(3) was detected from the hemolymph of reproductively active females by gas chromatography-mass spectrometry analysis while its titer in the hemolymph collected from diapausing adults was too low to be detected. These results demonstrated that JHSB(3) has biological function as a JH in P. stali. Topical application of JHSB(3), its stereoisomers and 10R-JH III also indicated that compounds with the 2R,3S-configuration were more potent than those with the 2S,3R-configuration and 2,3-double bond.
Sigma-Aldrich
Juvenile hormone III, ≥65%, liquid, non-sterile