Merck
  • Highly-oxygenated isopimarane-type diterpenes from Orthosiphon stamineus of Indonesia and their nitric oxide inhibitory activity.
Highly-oxygenated isopimarane-type diterpenes from Orthosiphon stamineus of Indonesia and their nitric oxide inhibitory activity.
Chemical & pharmaceutical bulletin (2003-03-04)
Suresh Awale, Yasuhiro Tezuka, Arjun H Banskota, I Ketut Adnyana, Shigetoshi Kadota
ABSTRACT
From the methanolic extract of Indonesian Orthosiphon stamineus, nine new highly-oxygenated isopimarane-type diterpenes [7-O-deacetylorthosiphol B (1), 6-hydroxyorthosiphol B (2), 3-O-deacetylorthosiphol I (3), 2-O-deacetylorthosiphol J (4), siphonols A-E (5-9)] have been isolated together with nine known diterpenes [orthosiphols H (10), K (11), M (12) and N (13); staminols A (14) and B (15); neoorthosiphols A (16) and B (17); norstaminol A (18)]. Their structures were determined based on the spectroscopic data. The isolated diterpenes inhibited nitric oxide (NO) production in lipopolysaccharide (LPS)-activated macrophage-like J774.1 cells. Compounds 4-7, 9, 10, 14, and 17 showed inhibitory activities more potent (IC(50), 10.8-25.5 microM) than a positive control N(G)-monomethyl-L-arginine (L-NMMA; IC(50), 26.0 microM).
Sigma-Aldrich
Isopimaric Acid, ≥98% (GC), powder