Merck
  • Endogenous Retroviruses Drive Lineage-Specific Regulatory Evolution across Primate and Rodent Placentae.
Endogenous Retroviruses Drive Lineage-Specific Regulatory Evolution across Primate and Rodent Placentae.
Molecular biology and evolution (2021-07-29)
Ming-An Sun, Gernot Wolf, Yejun Wang, Anna D Senft, Sherry Ralls, Jinpu Jin, Caitlin E Dunn-Fletcher, Louis J Muglia, Todd S Macfarlan
ABSTRACT
In mammals, the placenta mediates maternal-fetal nutrient and waste exchange and acts in an immunomodulatory way to facilitate maternal-fetal tolerance. The placenta is highly diverse across mammalian species, yet the molecular mechanisms that distinguish the placenta of human from other mammals are not fully understood. Using an interspecies transcriptomic comparison of human, macaque, and mouse late-gestation placentae, we identified hundreds of genes with lineage-specific expression-including dozens that are placentally enriched and potentially related to pregnancy. We further annotated the enhancers for different human tissues using epigenomic data and demonstrate that the placenta and chorion are unique in that their enhancers display the least conservation. We identified numerous lineage-specific human placental enhancers and found they highly overlap with specific families of endogenous retroviruses (ERVs), including MER21A, MER41A/B, and MER39B that were previously linked to immune response and placental function. Among these ERV families, we further demonstrate that MER41A/B insertions create dozens of lineage-specific serum response factor-binding loci in human, including one adjacent to FBN2, a placenta-specific gene with increased expression in humans that produces the peptide hormone placensin to stimulate glucose secretion and trophoblast invasion. Overall, our results demonstrate the prevalence of lineage-specific placental enhancers which are frequently associated with ERV insertions and likely facilitate the lineage-specific evolution of the mammalian placenta.
Sigma-Aldrich
TGF-β RI Kinase Inhibitor VI, SB431542, InSolution, ≥97%