- N-Alkylated aziridines are easily-prepared, potent, specific and cell-permeable covalent inhibitors of human β-glucocerebrosidase.
N-Alkylated aziridines are easily-prepared, potent, specific and cell-permeable covalent inhibitors of human β-glucocerebrosidase.
Chemical communications (Cambridge, England) (2015-06-19)
B T Adams, S Niccoli, M A Chowdhury, A N K Esarik, S J Lees, B P Rempel, C P Phenix
PMID26085065
ABSTRACT
β-Glucocerebrosidase deficiency leads to Gaucher disease and is a potential marker of Parkinson's disease. We have identified N-octyl conduritol aziridine as a potent and specific covalent inactivator of GBA1 in living cells. This compound is a promising lead towards a positron emission tomography probe intended to image GBA1 activity.