Merck
  • TBK1 and IKKε Act Redundantly to Mediate STING-Induced NF-κB Responses in Myeloid Cells.
TBK1 and IKKε Act Redundantly to Mediate STING-Induced NF-κB Responses in Myeloid Cells.
Cell reports (2020-04-09)
Katherine R Balka, Cynthia Louis, Tahnee L Saunders, Amber M Smith, Dale J Calleja, Damian B D'Silva, Fiona Moghaddas, Maximilien Tailler, Kate E Lawlor, Yifan Zhan, Christopher J Burns, Ian P Wicks, Jonathan J Miner, Benjamin T Kile, Seth L Masters, Dominic De Nardo
ABSTRACT
Stimulator of Interferon Genes (STING) is a critical component of host innate immune defense but can contribute to chronic autoimmune or autoinflammatory disease. Once activated, the cyclic guanosine monophosphate (GMP)-adenosine monophosphate (AMP) (cGAMP) synthase (cGAS)-STING pathway induces both type I interferon (IFN) expression and nuclear factor-κB (NF-κB)-mediated cytokine production. Currently, these two signaling arms are thought to be mediated by a single upstream kinase, TANK-binding kinase 1 (TBK1). Here, using genetic and pharmacological approaches, we show that TBK1 alone is dispensable for STING-induced NF-κB responses in human and mouse immune cells, as well as in vivo. We further demonstrate that TBK1 acts redundantly with IκB kinase ε (IKKε) to drive NF-κB upon STING activation. Interestingly, we show that activation of IFN regulatory factor 3 (IRF3) is highly dependent on TBK1 kinase activity, whereas NF-κB is significantly less sensitive to TBK1/IKKε kinase inhibition. Our work redefines signaling events downstream of cGAS-STING. Our findings further suggest that cGAS-STING will need to be targeted directly to effectively ameliorate the inflammation underpinning disorders associated with STING hyperactivity.
Sigma-Aldrich
5Z-7-Oxozeaenol, ≥98% (HPLC)
Sigma-Aldrich
TPCA-1, ≥95% (HPLC)
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from herring testes, Type XIV
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
DMXAA, ≥98% (HPLC), solid
Sigma-Aldrich
MRT67307 hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%