Merck
  • Discovery of a novel series of benzoic acid derivatives as potent and selective human beta3 adrenergic receptor agonists with good oral bioavailability. 3. Phenylethanolaminotetraline (PEAT) skeleton containing biphenyl or biphenyl ether moiety.
Discovery of a novel series of benzoic acid derivatives as potent and selective human beta3 adrenergic receptor agonists with good oral bioavailability. 3. Phenylethanolaminotetraline (PEAT) skeleton containing biphenyl or biphenyl ether moiety.
Journal of medicinal chemistry (2008-07-25)
Masashi Imanishi, Yutaka Nakajima, Yasuyo Tomishima, Hitoshi Hamashima, Kenichi Washizuka, Minoru Sakurai, Shigeo Matsui, Emiko Imamura, Koji Ueshima, Takao Yamamoto, Nobuhiro Yamamoto, Hirofumi Ishikawa, Keiko Nakano, Naoko Unami, Kaori Hamada, Yasuhiro Matsumura, Fujiko Takamura, Kouji Hattori
ABSTRACT
We designed a series of benzoic acid derivatives containing the biphenyl ether or biphenyl template on the RHS and a phenylethanolaminotetraline (PEAT) skeleton, which was prepared by highly stereoselective synthesis, to generate two structurally different lead compounds ( 10c, 10m) with a good balance of potency, selectivity, and pharmacokinetic profile. Further optimization of the two lead compounds to improve potency led to several potential candidates (i.e., 11f, 11l, 11o, 12b). In particular, biphenyl analogue 12b exhibited an excellent balance of high potency (EC50 = 0.38 nM) for beta3, high selectivity over beta1 and beta2, and good pharmacokinetic properties in rats, dogs, and monkeys.
Sigma-Aldrich
2-Amino-1-phenylethanol, 98%