Merck
  • Association between metabolic diseases and the culling risk of high-yielding dairy cows in a transition management facility using survival and decision tree analysis.
Association between metabolic diseases and the culling risk of high-yielding dairy cows in a transition management facility using survival and decision tree analysis.
Journal of dairy science (2018-08-21)
M Probo, O Bogado Pascottini, S LeBlanc, G Opsomer, M Hostens
ABSTRACT
The objective of this study was to assess the association between individual metabolic diseases (MD) and multiple MD (MD+) in the transition period (±3 wk relative to calving) and the culling risk in the first 120 d in milk (DIM) in Holstein-Friesian dairy cows. Health records from a transition management facility in Germany with 1,946 calvings were analyzed in a 1-yr cohort via survival analysis and a decision tree model. The recorded MD were milk fever (MF), retained placenta (RP), metritis (METR), ketosis (KET), displaced abomasum (DA), twinning (TWIN), and clinical mastitis (MAST). The overall culling within 120 DIM was 18%. The 120 DIM culling risk for healthy cows (64.8% of the total) was 13%, whereas it was 25% for MD (24.5%) and 33% for MD+ (10.7%) cows. The 120 DIM culling risk (%) for each MD and MD+, respectively, was 34.6 and 48 for MF and MF+, 15 and 31.5 for RP and RP+, 9.4 and 22.2 for METR and METR+, 30.7 and 37.3 for KET and KET+, 56.1 and 46.8 for DA and DA+, 30.3 and 34 for TWIN and TWIN+, and 36.6 and 27.8 for MAST and MAST+. Moreover, the incidence risk (%) for each MD and MD+, respectively, was 4 and 2.6 for MF and MF+, 1 and 2.8 for RP and RP+, 8.7 and 6 for METR and METR+, 4.5 and 6.1 for KET and KET+, 0.8 and 2.4 for DA and DA+, 1.7 and 2.7 for TWIN and TWIN+, and 3.6 and 1.8 for MAST and MAST+. Setting the healthy cows as the referent, the 120 DIM hazard ratios (HR) for culling were MD 2.1, MD+ 2.9, MF 3.3, MF+ 4.6, RP+ 2.7, METR+ 1.8, KET 2.6, KET+ 3.3, DA 5.5, DA+ 4.5, TWIN 2.8, TWIN+ 3.0, MAST 3.1, and MAST+ 2.3. According to both decision tree and random forest analyses, MF was the most significant disease influencing survival, followed by DA, MAST, METR, and TWIN. In conclusion, the presence of MD or MD+ during the transition period was associated with increased culling risk in the first 120 DIM. The culling hazard was greater when an MD was complicated with another MD. In this study performed in a well-managed large farm, uncomplicated cases of RP (HR = 1.2) and METR (HR = 0.7) did not have an influence on the 120 DIM culling risk. Interestingly, both decision tree and random forest analyses pointed to MF and DA as main culling reasons in the first 120 DIM in the present dairy herd.
Product Description
Supelco
Ceftiofur hydrochloride, VETRANAL®, analytical standard