Merck
  • Removal of sialic acid from bull sperm decreases motility and mucus penetration ability but increases zona pellucida binding and polyspermic penetration in vitro.
Removal of sialic acid from bull sperm decreases motility and mucus penetration ability but increases zona pellucida binding and polyspermic penetration in vitro.
Reproduction (Cambridge, England) (2018-04-06)
B Fernandez-Fuertes, A Blanco-Fernandez, C J Reid, K G Meade, S Fair, P Lonergan
ABSTRACT
This study tested the hypothesis that sperm sialic acid (Sia) is required to reach the site of fertilization, and that successful fertilization requires recognition of Sia from both the sperm and oocyte to occur. In addition, it has recently been reported that Siglecs (Sia-binding-immunoglobulin-like lectins) are present on the sperm surface. Thus, the possibility that the recognition of oocyte Sia was sperm-Siglec-mediated was also addressed. Sperm exposed to neuraminidase (NMase) exhibited lower overall and progressive motility, which translated to a decreased ability to swim through cervical mucus from cows in oestrus. In addition, when either sperm or cumulus-oocyte complexes (COCs) were treated with NMase, a decrease in cleavage and blastocyst rate was observed. However, incubation of sperm with increasing concentrations of anti-Siglec-2, -5, -6 and -10 antibodies prior to fertilization had no effect on their fertilizing ability. Interestingly, treatment with NMase increased the number of sperm bound to the ZP but also the rate of polyspermic fertilization. Flow cytometry analysis revealed no differences in the percentage of capacitated or acrosome-reacted sperm. These results suggest that Sia are required to reach the site of fertilization but need to be removed for sperm-oocyte interaction. However, fine regulation is needed to avoid abnormal fertilization which can lead to impaired embryo development.
Sigma-Aldrich
Merocyanine 540, Dye content 90 %