
Tricaproin

Synonym(s):
Glycerol trihexanoate, 1,2,3-Tricaproylglycerol, Glycerol tricapronate, Tricaproin, Trihexanoin
Linear Formula:
[CH3(CH2)4COOCH2]2CHOCO(CH2)4CH3
CAS Number:
Molecular Weight:
386.52
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing