
Rutaecarpine

Synonym(s):
8,13-Dihydroindolo[2′,3′:3,4]pyrido[2,1-b]quinazolin-5(7H)-one, Rhetine, Rutecarpine
Empirical Formula (Hill Notation):
C18H13N3O
CAS Number:
Molecular Weight:
287.32
Products
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing