
Repaglinide for system suitability

Synonym(s):
Repaglinide, (S)-(+)-2-Ethoxy-4-[N-[1-(2-piperidinophenyl)-3-methyl-1-butyl]aminocarbonylmethyl]benzoic acid, Novonorm, Prandin
Empirical Formula (Hill Notation):
C27H36N2O4
CAS Number:
Molecular Weight:
452.59
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing