
Palmitoyl-L-carnitine

Synonym(s):
(2R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxohexadecyl)oxy]-1-propanaminium inner salt, L-Carnitine hexadecanoyl ester, C16-Carnitine, Hexadecanoyl-L-carnitine
Empirical Formula (Hill Notation):
C23H45NO4
CAS Number:
Molecular Weight:
399.61
Products
Product Category
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing