
Octreotide impurity mixture

Synonym(s):
Octreotide, SMS 201-995
Empirical Formula (Hill Notation):
C49H66N10O10S2
CAS Number:
Molecular Weight:
1019.24
Products
Formula Weight
Gene Alias
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing