
Nilotinib System Suitability Mixture

Synonym(s):
4-Methyl-N-[3-(4-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl]-3-[[4-(3-pyridinyl)-2- pyrimidinyl]amino]-benzamide monohydrochloride monohydrate
Empirical Formula (Hill Notation):
C28H22F3N7O · HCl · H2O
CAS Number:
Molecular Weight:
583.99
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing