
Mepyramine maleate

Synonym(s):
Pyrilamine maleate salt, Mepyramine maleate salt, N-(4-Methoxyphenyl)methyl-N′,N′-dimethyl-N-(2-pyridinyl)-1,2-ethanediamine maleate salt
Empirical Formula (Hill Notation):
C17H23N3O · C4H4O4
CAS Number:
Molecular Weight:
401.46
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing