
Mepronil

Synonym(s):
3′-Isopropoxy-2-methylbenzanilide
Empirical Formula (Hill Notation):
C17H19NO2
CAS Number:
Molecular Weight:
269.34
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2