
L-α-Phosphatidylinositol ammonium salt from Glycine max (soybean)

Synonym(s):
1,2-Diacyl-sn-glycero-3-phospho-(1-D-myo-inositol), PI, PtdIns
Products
Product Category
Brand
Biological Source
Physical Form
Purity
Quality Segment
Functional Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing