
L-Alanine-13C3, d4

Linear Formula:
13CD313CD(NH2)13CO2H
Molecular Weight:
96.10
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing