
Furosemide for peak identification

Synonym(s):
Furosemide, 4-Chloro-N-furfuryl-5-sulfamoylanthranilic acid, 5-(Aminosulfonyl)-4-chloro-2-([2-furanylmethyl]amino)benzoic acid
Empirical Formula (Hill Notation):
C12H11ClN2O5S
CAS Number:
Molecular Weight:
330.74
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing