![Dexamethasone sodium phosphate for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/640/773/7f3f0723-f041-470e-8161-7c5b42846aff/640/7f3f0723-f041-470e-8161-7c5b42846aff.png)
Dexamethasone sodium phosphate for peak identification
![Dexamethasone sodium phosphate for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/640/773/7f3f0723-f041-470e-8161-7c5b42846aff/640/7f3f0723-f041-470e-8161-7c5b42846aff.png)
Synonym(s):
Dexamethasone 21-phosphate disodium salt
Empirical Formula (Hill Notation):
C22H28FNa2O8P
CAS Number:
Molecular Weight:
516.40
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing