
Cylindrospermopsin solution

Synonym(s):
6-[(R)-Hydroxy[(2aS,3R,4S,5aS,7R)-2,2a,3,4,5,5a,6,7-octahydro-3-methyl-4-(sulfooxy)-1H-1,8,8b-triazaacenaphthylen-7-yl]methyl]-2,4(1H,3H)-pyrimidinedione
Empirical Formula (Hill Notation):
C15H21N5O7S
CAS Number:
Molecular Weight:
415.42
Products
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing