
Butyl anthranilate

Linear Formula:
2-(H2N)C6H4CO2(CH2)3CH3
CAS Number:
Molecular Weight:
193.24
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing