
4,4′-Diformyltriphenylamine

Synonym(s):
4,4′-(Phenylimino)bisbenzaldehyde, Bis(4-formylphenyl)aniline, Bis(4-formylphenyl)phenylamine
Empirical Formula (Hill Notation):
C20H15NO2
CAS Number:
Molecular Weight:
301.34
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing