Merck
  • Folate-functionalized polymeric micelles for tumor targeted delivery of a potent multidrug-resistance modulator FG020326.
Folate-functionalized polymeric micelles for tumor targeted delivery of a potent multidrug-resistance modulator FG020326.
Journal of biomedical materials research. Part A (2007-10-18)
Xiaoqiang Yang, Wenjing Deng, Liwu Fu, Elvin Blanco, Jinming Gao, Daping Quan, Xintao Shuai
ABSTRACT
To overcome multidrug resistance (MDR) existing in tumor chemotherapy, polymeric micelles encoded with folic acid on the micelle surface were prepared with the encapsulation of a potent MDR modulator, FG020326. The micelles were fabricated from diblock copolymers of poly(ethylene glycol) (PEG) and biodegradable poly(epsilon-caprolactone) (PCL) with folate attached to the distal ends of PEG chains. The folate-conjugated copolymers, folate-PEG-PCL, were synthesized by multistep chemical reactions. First, allyl-terminated copolymer (allyl-PEG-PCL) was synthesized through a ring-opening polymerization of epsilon-caprolactone in bulk employing monoallyl-PEG as a macroinitiator. Second, the allyl terminal groups of copolymers were converted into primary amino groups by a radical addition reaction, followed by conjugation of the carboxylic group of folic acid. In vitro studies at 37 degrees C demonstrated that FG020326 release from micelles at pH 5.0 was faster than that at pH 7.4. Cytotoxicity studies with MTT assays indicated that folate-functionalized and FG020326-loaded micelles resensitized the cells approximately five times more than their folate-free counterparts (p < 0.01) in human KB(v200) cells treated with vincristine (VCR). The in vitro Rhodamine 123 efflux experiment using MDR KB(v200) cells revealed that when cells were pretreated with folate-attached and FG020326-loaded micelles, the P-glycoprotein (P-gp) drug efflux function was significantly inhibited.
Sigma-Aldrich
Amine-poly(ethylene glycol)-b-poly(ε-caprolactone), PEG average Mn 5,000, PCL average Mn 5,000