Merck
  • Single-isomer tetrasubstituted olefins from regioselective and stereospecific palladium-catalyzed coupling of beta-chloro-alpha-iodo-alpha,beta-unsaturated esters.
Single-isomer tetrasubstituted olefins from regioselective and stereospecific palladium-catalyzed coupling of beta-chloro-alpha-iodo-alpha,beta-unsaturated esters.
The Journal of organic chemistry (2006-04-22)
Alison B Lemay, Katarina S Vulic, William W Ogilvie
ABSTRACT
The efficient regioselective and stereospecific synthesis of tetrasubstituted olefins using a mild and convenient method is disclosed. 2-Alkynyl esters are selectively converted to E-beta-chloro-alpha-iodo-alpha,beta-unsaturated esters by exposure to Bu4NI in refluxing dichloroethane. These products are produced cleanly, regio- and stereoselectively, and in high yields. Single-isomer tetrasubstituted olefins bearing four different carbon substituents are then synthesized by sequential palladium-catalyzed coupling reactions. Selectivity results from reactivity differences in the intermediate substrates.
Sigma-Aldrich
Bis(triphenylphosphine)palladium(II) dichloride, 98%