Merck
  • Frequency of non-convulsive seizures and non-convulsive status epilepticus in subarachnoid hemorrhage patients in need of controlled ventilation and sedation.
Frequency of non-convulsive seizures and non-convulsive status epilepticus in subarachnoid hemorrhage patients in need of controlled ventilation and sedation.
Neurocritical care (2012-08-31)
Cecilia Lindgren, Erik Nordh, Silvana Naredi, Magnus Olivecrona
ABSTRACT
Non-convulsive seizures (NCSZ) can be more prevalent than previously recognized among comatose neuro-intensive care patients. The aim of this study was to evaluate the frequency of NCSZ and non-convulsive status epilepticus (NCSE) in sedated and ventilated subarachnoid hemorrhage (SAH) patients. Retrospective study at a university hospital neuro-intensive care unit, from January 2008 until June 2010. Patients were treated according to a local protocol, and were initially sedated with midazolam or propofol or combinations of these sedative agents. Thiopental was added for treatment of intracranial hypertension. No wake-up tests were performed. Using NicoletOne(®) equipment (VIASYS Healthcare Inc., USA), continuous EEG recordings based on four electrodes and a reference electrode was inspected at full length both in a two electrode bipolar and a four-channel referential montage. Approximately 5,500 h of continuous EEG were registered in 28 SAH patients (33 % of the patients eligible for inclusion). The median Glasgow Coma scale was 8 (range 3-14) and the median Hunt and Hess score was 4 (range 1-4). During EEG registration, no clinical seizures were observed. In none of the patients inter ictal epileptiform activity was seen. EEG seizures were recorded only in 2/28 (7 %) patients. One of the patients experienced 4 min of an NCSZ and one had a 5 h episode of an NCSE. Continuous EEG monitoring is important in detecting NCSZ in sedated patients. Continuous sedation, without wake-up tests, was associated with a low frequency of subclinical seizures in SAH patients in need of controlled ventilation.
Product Description
Supelco
(±)Thiopental solution, 1.0 mg/mL in methanol, analytical standard, for drug analysis
Sigma-Aldrich
(±)-Thiopental