Merck
  • An efficient, multicomponent approach for solvent-free synthesis of 2-amino-4H-chromene scaffold.
An efficient, multicomponent approach for solvent-free synthesis of 2-amino-4H-chromene scaffold.
Molecular diversity (2010-04-08)
M Reza Naimi-Jamal, Sara Mashkouri, Ali Sharifi
ABSTRACT
Solvent-free one-pot synthesis of 2-amino-4H-chromene scaffold is described in a very simple, efficient, and environmentally benign method using sodium carbonate as a cheap and non-toxic catalyst with up to excellent yields.
Sigma-Aldrich
4-Chlorobenzaldehyde, 97%