Merck
  • Analysis of the lidocaine metabolite 2,6-dimethylaniline in bovine and human milk.
Analysis of the lidocaine metabolite 2,6-dimethylaniline in bovine and human milk.
Journal of analytical toxicology (2002-01-05)
N W Puente, P D Josephy
ABSTRACT
2,6-Dimethylaniline (2,6-xylidine; 2,6-DMA) is a nasal carcinogen in rats. Humans may be exposed to this compound via several routes: 2,6-DMA is found in cigarette smoke; it is a pharmacologically inactive metabolite of some drugs (e.g., the local anesthetic lidocaine) and pesticides (e.g., metalaxyl); and it is an impurity in technical grade metalaxyl. The potential transfer of 2,6-DMA from mother to nursing infant via milk is of toxicological concern. Solid-phase microextraction with separation and detection using gas chromatography-mass spectrometry was optimized and used for the analysis of 2,6-DMA in milk. 2,6-DMA-d9 was synthesized and used for quantitation by the isotope ratio method. At a concentration of 5 ppb 2,6-DMA, the method detection limit was 0.20 ppb, and the relative standard deviation was 3.6%. Samples of milk were obtained from bovines administered lidocaine (2.9-3.9 mg/kg) during surgery. A breast milk sample was also obtained from a human donor who received 36 mg lidocaine during dental work. 2,6-DMA was present at levels ranging from 14.5 to 66.0 ppb in bovine milk and was detected at 1.6 ppb in the human milk sample. Our results demonstrate that 2,6-DMA, formed by the metabolism of lidocaine, is transferable to bovine and human milk.
Product Description
Supelco
2,6-Dimethylaniline, PESTANAL®, analytical standard
Sigma-Aldrich
2,6-Dimethylaniline, 99%
Supelco
2,6-Dimethylaniline, analytical standard