Merck
  • The human tumor microbiome is composed of tumor type-specific intracellular bacteria.
The human tumor microbiome is composed of tumor type-specific intracellular bacteria.
Science (New York, N.Y.) (2020-05-30)
Deborah Nejman, Ilana Livyatan, Garold Fuks, Nancy Gavert, Yaara Zwang, Leore T Geller, Aviva Rotter-Maskowitz, Roi Weiser, Giuseppe Mallel, Elinor Gigi, Arnon Meltser, Gavin M Douglas, Iris Kamer, Vancheswaran Gopalakrishnan, Tali Dadosh, Smadar Levin-Zaidman, Sofia Avnet, Tehila Atlan, Zachary A Cooper, Reetakshi Arora, Alexandria P Cogdill, Md Abdul Wadud Khan, Gabriel Ologun, Yuval Bussi, Adina Weinberger, Maya Lotan-Pompan, Ofra Golani, Gili Perry, Merav Rokah, Keren Bahar-Shany, Elisa A Rozeman, Christian U Blank, Anat Ronai, Ron Shaoul, Amnon Amit, Tatiana Dorfman, Ran Kremer, Zvi R Cohen, Sagi Harnof, Tali Siegal, Einav Yehuda-Shnaidman, Einav Nili Gal-Yam, Hagit Shapira, Nicola Baldini, Morgan G I Langille, Alon Ben-Nun, Bella Kaufman, Aviram Nissan, Talia Golan, Maya Dadiani, Keren Levanon, Jair Bar, Shlomit Yust-Katz, Iris Barshack, Daniel S Peeper, Dan J Raz, Eran Segal, Jennifer A Wargo, Judith Sandbank, Noam Shental, Ravid Straussman
ABSTRACT
Bacteria were first detected in human tumors more than 100 years ago, but the characterization of the tumor microbiome has remained challenging because of its low biomass. We undertook a comprehensive analysis of the tumor microbiome, studying 1526 tumors and their adjacent normal tissues across seven cancer types, including breast, lung, ovary, pancreas, melanoma, bone, and brain tumors. We found that each tumor type has a distinct microbiome composition and that breast cancer has a particularly rich and diverse microbiome. The intratumor bacteria are mostly intracellular and are present in both cancer and immune cells. We also noted correlations between intratumor bacteria or their predicted functions with tumor types and subtypes, patients' smoking status, and the response to immunotherapy.
Sigma-Aldrich
5R-Plex kit, Ultra-sensitive 16S NGS assay for degraded and low biomass DNA
Pricing and availability is not currently available.