Merck
  • Synthesis of scyllo-inositol derivatives and their effects on amyloid beta peptide aggregation.
Synthesis of scyllo-inositol derivatives and their effects on amyloid beta peptide aggregation.
Bioorganic & medicinal chemistry (2008-07-19)
Yedi Sun, Guohua Zhang, Cheryl A Hawkes, James E Shaw, JoAnne McLaurin, Mark Nitz
ABSTRACT
scyllo-Inositol has shown promise as a potential therapeutic for Alzheimer's disease, by directly interacting with the amyloid beta (Abeta) peptide to inhibit Abeta42 fiber formation. To explore the molecular details of the inositol-Abeta42 interaction, a series of scyllo-inositol derivatives have been synthesized which contain deoxy, fluoro, chloro, and methoxy substitutions. The effects of these compounds on the aggregation cascade of Abeta42 have been investigated using electron microscopy (EM). EM analyses revealed that the 1-deoxy-1-fluoro- and 1,4-dimethyl-scyllo-inositols significantly inhibit the formation of Abeta42 fibers. The other derivatives showed some alterations in the morphology of the Abeta42 fibers produced. These findings indicate the importance of all of the hydroxyl groups of scyllo-inositol for complete inhibition of Abeta aggregation.
Sigma-Aldrich
O-Phenyl chlorothionoformate, 99%
Sigma-Aldrich
scyllo-Inositol, ≥98%