Merck
  • Condensed Chromatin Behaves like a Solid on the Mesoscale In Vitro and in Living Cells.
Condensed Chromatin Behaves like a Solid on the Mesoscale In Vitro and in Living Cells.
Cell (2020-12-17)
Hilmar Strickfaden, Thomas O Tolsma, Ajit Sharma, D Alan Underhill, Jeffrey C Hansen, Michael J Hendzel
ABSTRACT
The association of nuclear DNA with histones to form chromatin is essential for temporal and spatial control of eukaryotic genomes. In this study, we examined the physical state of condensed chromatin in vitro and in vivo. Our in vitro studies demonstrate that self-association of nucleosomal arrays under a wide range of solution conditions produces supramolecular condensates in which the chromatin is physically constrained and solid-like. By measuring DNA mobility in living cells, we show that condensed chromatin also exhibits solid-like behavior in vivo. Representative heterochromatin proteins, however, display liquid-like behavior and coalesce around the solid chromatin scaffold. Importantly, euchromatin and heterochromatin show solid-like behavior even under conditions that produce limited interactions between chromatin fibers. Our results reveal that condensed chromatin exists in a solid-like state whose properties resist external forces and create an elastic gel and provides a scaffold that supports liquid-liquid phase separation of chromatin binding proteins.
Sigma-Aldrich
Aminoallyl-dUTP sodium salt, ≥85%, lyophilized powder
Sigma-Aldrich
Maleimide, 99%
Sigma-Aldrich
5-Carboxy-tetramethylrhodamine N-succinimidyl ester, BioReagent, suitable for fluorescence
Sigma-Aldrich
(-)-JQ1, ≥95% (HPLC)