Merck
  • Protective effects of Erdosteine on interleukin-1β-stimulated inflammation via inhibiting the activation of MAPK, NF-κB, and Wnt/β-catenin signaling pathways in rat osteoarthritis.
Protective effects of Erdosteine on interleukin-1β-stimulated inflammation via inhibiting the activation of MAPK, NF-κB, and Wnt/β-catenin signaling pathways in rat osteoarthritis.
European journal of pharmacology (2020-01-21)
Yang Xi, Xiaojian Huang, Genmei Tan, Xiangyu Chu, Rui Zhang, Xiaohu Ma, Bowei Ni, Hongbo You
ABSTRACT
Osteoarthritis (OA), a degenerative arthropathy, is featured with progressive degradation of cartilage and a chondrocyte inflammatory response. Erdosteine (ER) showed the anti-oxidant properties and various anti-inflammatory effects in various diseases. However, whether it protects against OA remains unknown. In this study, we explore the potential therapeutic properties of ER on IL-1β-stimulated rat chondrocytes and its underlying mechanism in vitro and vivo. Cell viability, pro-inflammatory cytokines and the degradation of ECM biomarkers were tested to determine the effects of ER at 10, 20, and 40 μM doses on IL-1β-induced rat chondrocytes for 24 h in virto. In vivo, intra-articular injections of 50 μl of 100 mg/ml ER twice a week for 8 weeks. The results showed ER significantly suppressed the expressions of IL-1β-induced the production of inflammatory factors in a dose-dependent pattern (4.30-fold decrease in COX-2, p < 0.05; 4.77-fold decrease in iNOS, p < 0.05 at 40 μM in protein levels). Moreover, ER could attenuate the degradation of ECM in IL-1β-induced rat chondrocytes by repressing the expression of OA-related factors (2.40-fold decrease in ADAMTS-5, p < 0.05; 3.12-fold decrease in MMP1, p < 0.05; 3.97-fold decrease in MMP3, p < 0.05; and 2.62-fold decrease in MMP-13, p < 0.05 at 40 μM in protein levels). Furthermore, our study revealed that ER could inhibit the activations of IL-1β-induced MAPK and Wnt/β-catenin. Besides, ER could suppress the process of IL-1β-induced P65 from the cytoplasm into the nucleus. In vivo, ER delaied the osteoarthritis progression in rat OA models. Collectively, ER might become a new therapeutic agent for OA.
Sigma-Aldrich
Erdosteine, ≥98% (HPLC)
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder