Merck
  • A protective human monoclonal antibody targeting the West Nile virus E protein preferentially recognizes mature virions.
A protective human monoclonal antibody targeting the West Nile virus E protein preferentially recognizes mature virions.
Nature microbiology (2018-11-21)
Leslie Goo, Kari Debbink, Nurgun Kose, Gopal Sapparapu, Michael P Doyle, Alex W Wessel, Justin M Richner, Katherine E Burgomaster, Bridget C Larman, Kimberly A Dowd, Michael S Diamond, James E Crowe, Theodore C Pierson
ABSTRACT
West Nile virus (WNV), a member of the Flavivirus genus, is a leading cause of viral encephalitis in the United States1. The development of neutralizing antibodies against the flavivirus envelope (E) protein is critical for immunity and vaccine protection2. Previously identified candidate therapeutic mouse and human neutralizing monoclonal antibodies (mAbs) target epitopes within the E domain III lateral ridge and the domain I-II hinge region, respectively3. To explore the neutralizing antibody repertoire elicited by WNV infection for potential therapeutic application, we isolated ten mAbs from WNV-infected individuals. mAb WNV-86 neutralized WNV with a 50% inhibitory concentration of 2 ng ml-1, one of the most potently neutralizing flavivirus-specific antibodies ever isolated. WNV-86 targets an epitope in E domain II, and preferentially recognizes mature virions lacking an uncleaved form of the chaperone protein prM, unlike most flavivirus-specific antibodies4. In vitro selection experiments revealed a neutralization escape mechanism involving a glycan addition to E domain II. Finally, a single dose of WNV-86 administered two days post-infection protected mice from lethal WNV challenge. This study identifies a highly potent human neutralizing mAb with therapeutic potential that targets an epitope preferentially displayed on mature virions.
Sigma-Aldrich
Ouabain octahydrate, ≥95% (HPLC), powder
Sigma-Aldrich
Anti-Mouse IgG (H+L), Biotin antibody produced in goat, ~2 mg/mL, affinity isolated antibody