Merck
  • Genetically encoded optical activation of DNA recombination in human cells.
Genetically encoded optical activation of DNA recombination in human cells.
Chemical communications (Cambridge, England) (2016-06-10)
J Luo, E Arbely, J Zhang, C Chou, R Uprety, J W Chin, A Deiters
ABSTRACT
We developed two tightly regulated, light-activated Cre recombinase enzymes through site-specific incorporation of two genetically-encoded photocaged amino acids in human cells. Excellent optical off to on switching of DNA recombination was achieved. Furthermore, we demonstrated precise spatial control of Cre recombinase through patterned illumination.
Sigma-Aldrich
Methyl-o-nitropiperonyllysine, ≥95%