
Tetrakis(ethylmethylamido)hafnium(IV)

Synonym(s):
TEMAH, Tetrakis(ethylmethylamino)hafnium(IV)
Linear Formula:
[(CH3)(C2H5)N]4Hf
CAS Number:
Molecular Weight:
410.90
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing