
Prolylphenylalanine

Synonym(s):
Pro-Phe
Empirical Formula (Hill Notation):
C14H18N2O3
CAS Number:
Molecular Weight:
262.30
Products
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing