
Josamycin for peak identification

Synonym(s):
Josamycin
Empirical Formula (Hill Notation):
C42H69NO15
CAS Number:
Molecular Weight:
827.99
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing