
Forsythoside A

Synonym(s):
Forsythiaside
Empirical Formula (Hill Notation):
C29H36O15
CAS Number:
Molecular Weight:
624.59
Products
Formula Weight
Manufacturer
Product Line
Format
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing