
di-Lithium tetraborate

Synonym(s):
di-Lithium tetraborate
Empirical Formula (Hill Notation):
B4Li2O7
CAS Number:
Molecular Weight:
169.12
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing