
D-Galactose-1-d

Empirical Formula (Hill Notation):
C6DH11O6
CAS Number:
Molecular Weight:
181.16
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Showing 1-2 of 2
Compare
Product Number
Description
Pricing