
Cyclophosphamide Related Compound C

Synonym(s):
3-Aminopropyl monophosphate
Empirical Formula (Hill Notation):
C3H10NO4P
CAS Number:
Molecular Weight:
155.09
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing