
Cilastatin sodium

Synonym(s):
Cilastatin sodium salt
Empirical Formula (Hill Notation):
C16H25N2NaO5S
CAS Number:
Molecular Weight:
380.43
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing