Skip to Content
Merck
Capsaicin European Pharmacopoeia (EP) Reference Standard

Capsaicin

Capsaicin European Pharmacopoeia (EP) Reference Standard
Synonym(s):
8-Methyl-N-vanillyl-trans-6-nonenamide, Capsaicin
Linear Formula:
(CH3)2CHCH=CH(CH2)4CONHCH2C6H3-4-(OH)-3-(OCH3)
CAS Number:
Molecular Weight:
305.41
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Research Area
Chemical Composition
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique