
Calcium glucoheptonate

Synonym(s):
Calcium gluceptate, α-D-Glucoheptonic acid calcium salt, D-Glycero-D-glucoheptonic acid calcium salt, Calcium α-D-heptagluconate
Empirical Formula (Hill Notation):
C14H26CaO16
CAS Number:
Molecular Weight:
490.42
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing